Product Details

SNP ID
rs140299857
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:62196448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGAATTCTAGGCACCAAGGCACA[G/T]CTTAGAGTAGACCCGAGTCCTGCTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
L1TD1 PubMed Links

Gene Details

Gene
L1TD1
Gene Name
LINE1 type transposase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details