Product Details

SNP ID
rs140577325
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:43653205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGAGTCTGAAACTCTGAATCC[C/T]AGTGCTAGGATAATGACCTTTTATC
Phenotype
MIM: 609764
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM4A PubMed Links

Gene Details

Gene
KDM4A
Gene Name
lysine demethylase 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014663.2 233 Silent Mutation CCC,CCT P10P NP_055478.2
XM_005271354.3 233 Silent Mutation CCC,CCT P10P XP_005271411.1
XM_005271355.3 233 Silent Mutation CCC,CCT P10P XP_005271412.1
XM_005271356.3 233 UTR 5 XP_005271413.1
XM_011542467.2 233 Intron XP_011540769.2
XM_017002912.1 233 UTR 5 XP_016858401.1

View Full Product Details