Product Details

SNP ID
rs141334808
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:29121280 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGGTCCCAGATTTCTGGCTCTGC[A/G]CAGCCTACGGCTCGGGGACTCCTAG
Phenotype
MIM: 130500
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPB41 PubMed Links

Gene Details

Gene
EPB41
Gene Name
erythrocyte membrane protein band 4.1
There are no transcripts associated with this gene.

Gene
TMEM200B
Gene Name
transmembrane protein 200B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003682.3 623 Silent Mutation TGC,TGT C183C NP_001003682.1
NM_001171868.1 623 Silent Mutation TGC,TGT C183C NP_001165339.1
XM_011541457.2 623 Silent Mutation TGC,TGT C194C XP_011539759.1

View Full Product Details