Product Details

SNP ID
rs142474066
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:66798134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAGCTCTTCAGTGGGCATCTGT[C/G]CACCCCAAAGACGGTCTTCCCCTGA
Phenotype
MIM: 606413
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
INSL5 PubMed Links

Gene Details

Gene
INSL5
Gene Name
insulin like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005478.4 325 Missense Mutation GCA,GGA A96G NP_005469.2

View Full Product Details