Product Details

SNP ID
rs142514473
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:158398923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCTAACTTGATAACAGGTGCCA[C/T]GTCACAGAAATAGTGGTTAACCCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10T2 PubMed Links

Gene Details

Gene
OR10T2
Gene Name
olfactory receptor family 10 subfamily T member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004475.1 544 Missense Mutation ATG,GTG M182V NP_001004475.1

View Full Product Details