Product Details

SNP ID
rs143105287
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:158398885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTAAATAAAGCCAGCTCTTTCAC[A/G]TGGGTGTCAGTGCAGGCTAACTTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10T2 PubMed Links

Gene Details

Gene
OR10T2
Gene Name
olfactory receptor family 10 subfamily T member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004475.1 582 Silent Mutation CAC,CAT H194H NP_001004475.1

View Full Product Details