Product Details

SNP ID
rs143745964
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:209675899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGAGCTGCAGGCCGCCCTGGAGC[A/G]ACAGGCTCTCCAGAAGCAAGCCCTG
Phenotype
MIM: 614447
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
G0S2 PubMed Links

Gene Details

Gene
G0S2
Gene Name
G0/G1 switch 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015714.3 472 Missense Mutation CAA,CGA Q72R NP_056529.1
Gene
LOC101930114
Gene Name
uncharacterized LOC101930114
There are no transcripts associated with this gene.

View Full Product Details