Product Details

SNP ID
rs144925228
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:29120913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGTCTCCCCTCTCTTCAGACCC[A/G]GGCCCCCAAGTCCCCTCCTCCTCCC
Phenotype
MIM: 130500
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPB41 PubMed Links

Gene Details

Gene
EPB41
Gene Name
erythrocyte membrane protein band 4.1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166005.1 990 Intron NP_001159477.1
NM_001166006.1 990 Intron NP_001159478.1
NM_001166007.1 990 Intron NP_001159479.1
NM_004437.3 990 Intron NP_004428.1
NM_203342.2 990 Intron NP_976217.1
NM_203343.2 990 Intron NP_976218.1
XM_005245753.1 990 Intron XP_005245810.1
XM_005245757.1 990 Intron XP_005245814.1
XM_005245760.1 990 Intron XP_005245817.1
XM_005245761.1 990 Intron XP_005245818.1
XM_005245763.1 990 Intron XP_005245820.1
XM_005245764.1 990 Intron XP_005245821.1
XM_005245765.1 990 Intron XP_005245822.1
XM_005245768.1 990 Intron XP_005245825.1
XM_005245769.1 990 Intron XP_005245826.1
XM_005245770.1 990 Intron XP_005245827.1
XM_005245772.4 990 Intron XP_005245829.1
XM_005245773.4 990 Intron XP_005245830.1
XM_005245774.1 990 Intron XP_005245831.1
XM_006710434.1 990 Intron XP_006710497.1
XM_006710439.1 990 Intron XP_006710502.1
XM_011540956.1 990 Intron XP_011539258.1
XM_011540957.1 990 Intron XP_011539259.1
XM_011540958.1 990 Intron XP_011539260.1
XM_011540959.1 990 Intron XP_011539261.1
XM_011540960.1 990 Intron XP_011539262.1
XM_011540961.1 990 Intron XP_011539263.1
XM_011540962.1 990 Intron XP_011539264.1
XM_011540963.2 990 Intron XP_011539265.1
XM_011540964.1 990 Intron XP_011539266.1
XM_011540965.2 990 Intron XP_011539267.1
XM_017000581.1 990 Intron XP_016856070.1
XM_017000582.1 990 Intron XP_016856071.1
XM_017000583.1 990 Intron XP_016856072.1
XM_017000584.1 990 Intron XP_016856073.1
XM_017000585.1 990 Intron XP_016856074.1
XM_017000586.1 990 Intron XP_016856075.1
XM_017000587.1 990 Intron XP_016856076.1
XM_017000588.1 990 Intron XP_016856077.1
XM_017000589.1 990 Intron XP_016856078.1
XM_017000590.1 990 Intron XP_016856079.1
XM_017000591.1 990 Intron XP_016856080.1
XM_017000592.1 990 Intron XP_016856081.1
XM_017000593.1 990 Intron XP_016856082.1
XM_017000594.1 990 Intron XP_016856083.1
XM_017000595.1 990 Intron XP_016856084.1
XM_017000596.1 990 Intron XP_016856085.1
XM_017000597.1 990 Intron XP_016856086.1
XM_017000598.1 990 Intron XP_016856087.1
XM_017000599.1 990 Intron XP_016856088.1
XM_017000600.1 990 Intron XP_016856089.1
XM_017000601.1 990 Intron XP_016856090.1
XM_017000602.1 990 Intron XP_016856091.1
XM_017000603.1 990 Intron XP_016856092.1
XM_017000604.1 990 Intron XP_016856093.1
Gene
TMEM200B
Gene Name
transmembrane protein 200B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003682.3 990 Missense Mutation CGG,TGG R306W NP_001003682.1
NM_001171868.1 990 Missense Mutation CGG,TGG R306W NP_001165339.1
XM_011541457.2 990 Missense Mutation CGG,TGG R317W XP_011539759.1

View Full Product Details