Product Details

SNP ID
rs145645025
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:1243503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACACGGGGGCCGTGAACCGACCC[A/G]AGGCCAGGCTCAGACCGGAGCCTCG
Phenotype
MIM: 615291 MIM: 616593
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B3GALT6 PubMed Links

Gene Details

Gene
B3GALT6
Gene Name
beta-1,3-galactosyltransferase 6
There are no transcripts associated with this gene.

Gene
FAM132A
Gene Name
family with sequence similarity 132 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014980.2 711 Missense Mutation TCG,TTG S194L NP_001014980.1
XM_011541435.2 711 Missense Mutation TCG,TTG S194L XP_011539737.1
XM_011541436.2 711 Missense Mutation TCG,TTG S194L XP_011539738.1
XM_011541437.1 711 Missense Mutation TCG,TTG S122L XP_011539739.1

View Full Product Details