Product Details

SNP ID
rs145813190
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:74738719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTCTGAAGAAAGCAAATGGTGA[A/T]GCCACTTTGAAATTTGAACCATTTG
Phenotype
MIM: 123691 MIM: 611245
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CRYZ PubMed Links

Gene Details

Gene
CRYZ
Gene Name
crystallin zeta
There are no transcripts associated with this gene.

Gene
TYW3
Gene Name
tRNA-yW synthesizing protein 3 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162916.1 374 Intron NP_001156388.1
NM_138467.2 374 Missense Mutation GAA,GAT E95D NP_612476.1
XM_006710347.1 374 Missense Mutation GAA,GAT E95D XP_006710410.1

View Full Product Details