Product Details

SNP ID
rs150276430
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:158480049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTTTCTTTTTCTGTATCTAGTC[A/G]TTCTTAGTGGCAATGTCACCATTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10R2 PubMed Links

Gene Details

Gene
OR10R2
Gene Name
olfactory receptor family 10 subfamily R member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004472.1 172 Missense Mutation ATT,GTT I58V NP_001004472.1

View Full Product Details