Product Details

SNP ID
rs139105685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2483167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATACACAGGTTTCTCCCGTCAGTG[C/T]GTCCTCTGGTGTCTGATGAGATTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF343 PubMed Links

Gene Details

Gene
ZNF343
Gene Name
zinc finger protein 343
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282495.1 2146 Silent Mutation ACA,ACG T598T NP_001269424.1
NM_001282496.1 2146 Silent Mutation ACA,ACG T598T NP_001269425.1
NM_001282497.1 2146 Silent Mutation ACA,ACG T639T NP_001269426.1
NM_001282498.1 2146 Silent Mutation ACA,ACG T508T NP_001269427.1
NM_001282499.1 2146 Intron NP_001269428.1
NM_001321800.1 2146 Silent Mutation ACA,ACG T598T NP_001308729.1
NM_001321801.1 2146 Silent Mutation ACA,ACG T639T NP_001308730.1
NM_001321802.1 2146 Silent Mutation ACA,ACG T638T NP_001308731.1
NM_001321803.1 2146 Silent Mutation ACA,ACG T638T NP_001308732.1
NM_001321805.1 2146 Silent Mutation ACA,ACG T597T NP_001308734.1
NM_024325.5 2146 Silent Mutation ACA,ACG T598T NP_077301.4
XM_017028062.1 2146 Silent Mutation ACA,ACG T598T XP_016883551.1

View Full Product Details