Product Details

SNP ID
rs143371176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:64073949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGTAGGAGTCCCGGTGCATGAG[C/T]GTGTAGATCTGCAGCTGCGCGTCGT
Phenotype
MIM: 602548 MIM: 605071 MIM: 604784
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LKAAEAR1 PubMed Links

Gene Details

Gene
LKAAEAR1
Gene Name
LKAAEAR motif containing 1
There are no transcripts associated with this gene.

Gene
MIR6813
Gene Name
microRNA 6813
There are no transcripts associated with this gene.

Gene
OPRL1
Gene Name
opioid related nociceptin receptor 1
There are no transcripts associated with this gene.

Gene
RGS19
Gene Name
regulator of G-protein signaling 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039467.1 825 Silent Mutation ACA,ACG T186T NP_001034556.1
NM_005873.2 825 Silent Mutation ACA,ACG T186T NP_005864.1
XM_005260182.3 825 Silent Mutation ACA,ACG T164T XP_005260239.1
XM_005260183.2 825 Silent Mutation ACA,ACG T164T XP_005260240.1
XM_011528484.1 825 Silent Mutation ACA,ACG T215T XP_011526786.1
XM_011528485.2 825 Silent Mutation ACA,ACG T186T XP_011526787.1
XM_011528486.2 825 Silent Mutation ACA,ACG T186T XP_011526788.1
Gene
TCEA2
Gene Name
transcription elongation factor A2
There are no transcripts associated with this gene.

View Full Product Details