Product Details

SNP ID
rs139868615
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:25726999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAGCACTGTCCAATAGACCTTTT[G/T]GTGATGTTAGAAACGCTCTCTGTCT
Phenotype
MIM: 603152 MIM: 600609 MIM: 606870
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ATP5J PubMed Links

Gene Details

Gene
ATP5J
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit F6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003696.1 Intron NP_001003696.1
NM_001003697.1 Intron NP_001003697.1
NM_001003701.1 Intron NP_001003701.1
NM_001003703.1 Intron NP_001003703.1
NM_001320266.1 Intron NP_001307195.1
NM_001320267.1 Intron NP_001307196.1
NM_001685.4 Intron NP_001676.2
Gene
GABPA
Gene Name
GA binding protein transcription factor alpha subunit
There are no transcripts associated with this gene.

Gene
JAM2
Gene Name
junctional adhesion molecule 2
There are no transcripts associated with this gene.

View Full Product Details