Product Details

SNP ID
rs147499380
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31168367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTACTTCACCTAACATGATGTT[C/T]TTGAGGTTCATTTATGTTGTAGCAT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MIR3928 PubMed Links

Gene Details

Gene
MIR3928
Gene Name
microRNA 3928
There are no transcripts associated with this gene.

Gene
RNF185
Gene Name
ring finger protein 185
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135825.1 Intron NP_001129297.1
NM_152267.3 Intron NP_689480.2

View Full Product Details