Product Details
- SNP ID
-
rs137853021
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:44942443 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGAGCGGCTGGGCCGCTTCCTCTG[G/T]TCGCTGCCCGTGGCCCCCGGGGCGT
- Phenotype
-
MIM: 603714
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SIX3
PubMed Links
Gene Details
- Gene
- SIX3
- Gene Name
- SIX homeobox 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005413.3 |
546 |
Missense Mutation |
TGG,TGT |
W113C |
NP_005404.1 |
- Gene
- SIX3-AS1
- Gene Name
- SIX3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details