Product Details

SNP ID
rs140876587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:221419052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAATAAATGGTTCAGAATCTATT[C/T]TTTTTCCTCATTTTTCCCTGATTTG
Phenotype
MIM: 602188
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPHA4 PubMed Links

Gene Details

Gene
EPHA4
Gene Name
EPH receptor A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304536.1 5391 UTR 3 NP_001291465.1
NM_001304537.1 5391 UTR 3 NP_001291466.1
NM_004438.4 5391 UTR 3 NP_004429.1
XM_005246374.2 5391 UTR 3 XP_005246431.1

View Full Product Details