Product Details

SNP ID
rs141939363
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:105241898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGTCCCTTGAGGCTTACACATAC[C/T]TGCTGCTGAACACCAGCAACGCCTC
Phenotype
MIM: 604838 MIM: 606237
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GPR45 PubMed Links

Gene Details

Gene
GPR45
Gene Name
G protein-coupled receptor 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007227.3 156 Silent Mutation CTG,TTG L14L NP_009158.3
Gene
TGFBRAP1
Gene Name
transforming growth factor beta receptor associated protein 1
There are no transcripts associated with this gene.

View Full Product Details