Product Details

SNP ID
rs148569674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:100273747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACACCAGCTTATTTATTCAAATT[C/T]GCTCTTCTTATTAAACAGAAAAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LONRF2 PubMed Links

Gene Details

Gene
LONRF2
Gene Name
LON peptidase N-terminal domain and ring finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198461.3 13456 UTR 3 NP_940863.3
XM_017003491.1 13456 Intron XP_016858980.1

View Full Product Details