Product Details

SNP ID
rs149156170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176151731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGCGGCTACCTAGGCGAGCAGG[A/G]CGCCGACTACTACGGCGGCGGCGCG
Phenotype
MIM: 142980 MIM: 142981 MIM: 611576
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOXD3 PubMed Links

Gene Details

Gene
HOXD3
Gene Name
homeobox D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006898.4 1338 Intron NP_008829.3
XM_005246509.3 1338 Intron XP_005246566.1
XM_005246510.4 1338 Intron XP_005246567.1
XM_005246511.3 1338 Intron XP_005246568.1
XM_005246513.4 1338 Intron XP_005246570.1
XM_006712477.2 1338 Intron XP_006712540.1
XM_011511065.2 1338 Intron XP_011509367.1
XM_011511066.2 1338 Intron XP_011509368.1
Gene
HOXD4
Gene Name
homeobox D4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014621.2 1338 Missense Mutation GAC,GGC D33G NP_055436.2
XM_005246514.4 1338 Missense Mutation GAC,GGC D33G XP_005246571.1
Gene
MIR10B
Gene Name
microRNA 10b
There are no transcripts associated with this gene.

View Full Product Details