Product Details

SNP ID
rs139699451
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50607722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCGGCACAGGTGTTGCAGGCTG[C/T]GGGCACTGCTTCTGCGTACAAAGGG
Phenotype
MIM: 602441 MIM: 602130
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CISH PubMed Links
Additional Information
For this assay, SNP(s) [rs2239753] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CISH
Gene Name
cytokine inducible SH2 containing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013324.5 3665 Missense Mutation NP_037456.5
NM_145071.2 3665 Missense Mutation NP_659508.1
XM_011533329.2 3665 Missense Mutation XP_011531631.1
Gene
MAPKAPK3
Gene Name
mitogen-activated protein kinase-activated protein kinase 3
There are no transcripts associated with this gene.

View Full Product Details