Product Details

SNP ID
rs146701862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108908075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTGTCAGGAGATTTGGAGGAGT[C/T]TGTCTCCATGTCTGGCTGCTTCCCA
Phenotype
MIM: 605128
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCA1C PubMed Links

Gene Details

Gene
GUCA1C
Gene Name
guanylate cyclase activator 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005459.3 645 Missense Mutation AAC,GAC N193D NP_005450.3
XM_005247910.3 645 UTR 3 XP_005247967.1
XM_011513334.2 645 Missense Mutation AAC,GAC N109D XP_011511636.1

View Full Product Details