Product Details

SNP ID
rs149789249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45394650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTTGAGAAATGGTGGCATCAAC[A/G]AATAAAAGAACAGGCCTCCAAAATT
Phenotype
MIM: 604544
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LARS2 PubMed Links

Gene Details

Gene
LARS2
Gene Name
leucyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015340.3 382 Missense Mutation CAA,CGA Q66R NP_056155.1
XM_005265006.1 382 Missense Mutation CAA,CGA Q66R XP_005265063.1
XM_011533554.2 382 Missense Mutation CAA,CGA Q66R XP_011531856.1
XM_017006042.1 382 Missense Mutation CAA,CGA Q66R XP_016861531.1

View Full Product Details