Product Details

SNP ID
rs142074768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:121761675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGAATCGCACCAGATGGCATCAG[A/T]TCTGCATCTACAGCTGTGCCCTAAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PP12613 PubMed Links

Gene Details

Gene
PP12613
Gene Name
uncharacterized LOC100192379
There are no transcripts associated with this gene.

Gene
TMEM155
Gene Name
transmembrane protein 155
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317837.1 612 Missense Mutation GAA,GAT E25D NP_001304766.1
NM_001317838.1 612 Missense Mutation GAA,GAT E25D NP_001304767.1
NM_001317839.1 612 Missense Mutation GAA,GAT E25D NP_001304768.1
NM_001317841.1 612 UTR 5 NP_001304770.1
NM_001317842.1 612 UTR 5 NP_001304771.1
NM_152399.3 612 Missense Mutation GAA,GAT E25D NP_689612.3
XM_005262737.2 612 Missense Mutation GAA,GAT E25D XP_005262794.1
XM_017007725.1 612 Missense Mutation GAA,GAT E25D XP_016863214.1
XM_017007726.1 612 UTR 5 XP_016863215.1

View Full Product Details