Product Details

SNP ID
rs149760380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:121760416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGGATTTGAACTGGCATGCCCC[A/G]TTTTCTCTGTGGGCTTCCTCCTTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PP12613 PubMed Links

Gene Details

Gene
PP12613
Gene Name
uncharacterized LOC100192379
There are no transcripts associated with this gene.

Gene
TMEM155
Gene Name
transmembrane protein 155
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317837.1 808 Missense Mutation CGG,TGG R91W NP_001304766.1
NM_001317838.1 808 Missense Mutation CGG,TGG R91W NP_001304767.1
NM_001317839.1 808 Missense Mutation CGG,TGG R91W NP_001304768.1
NM_001317841.1 808 Missense Mutation CGG,TGG R65W NP_001304770.1
NM_001317842.1 808 Missense Mutation CGG,TGG R65W NP_001304771.1
NM_152399.3 808 Missense Mutation CGG,TGG R91W NP_689612.3
XM_005262737.2 808 Missense Mutation CGG,TGG R91W XP_005262794.1
XM_017007725.1 808 Missense Mutation CGG,TGG R91W XP_016863214.1
XM_017007726.1 808 Missense Mutation CGG,TGG R65W XP_016863215.1

View Full Product Details