Product Details
- SNP ID
-
rs151159930
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:84677377 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAACTCGGCCTCGACTGTCACCAA[C/T]GAGGATCCTACTGTGATCCCTGCAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WDFY3
PubMed Links
Gene Details
- Gene
- WDFY3
- Gene Name
- WD repeat and FYVE domain containing 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014991.4 |
10639 |
Missense Mutation |
ATT,GTT |
I3427V |
NP_055806.2 |
XM_005262858.4 |
10639 |
Missense Mutation |
ATT,GTT |
I3445V |
XP_005262915.1 |
XM_011531757.2 |
10639 |
Missense Mutation |
ATT,GTT |
I3445V |
XP_011530059.1 |
XM_011531759.2 |
10639 |
Missense Mutation |
ATT,GTT |
I3445V |
XP_011530061.1 |
XM_011531760.2 |
10639 |
Missense Mutation |
ATT,GTT |
I3445V |
XP_011530062.1 |
XM_011531761.2 |
10639 |
Missense Mutation |
ATT,GTT |
I3430V |
XP_011530063.1 |
XM_011531762.2 |
10639 |
Missense Mutation |
ATT,GTT |
I3428V |
XP_011530064.1 |
XM_011531763.2 |
10639 |
Missense Mutation |
ATT,GTT |
I3427V |
XP_011530065.1 |
XM_011531764.2 |
10639 |
Missense Mutation |
ATT,GTT |
I3417V |
XP_011530066.1 |
XM_011531765.2 |
10639 |
Missense Mutation |
ATT,GTT |
I3410V |
XP_011530067.1 |
XM_011531766.2 |
10639 |
Intron |
|
|
XP_011530068.1 |
XM_011531767.2 |
10639 |
Intron |
|
|
XP_011530069.1 |
XM_017007906.1 |
10639 |
Missense Mutation |
ATT,GTT |
I3445V |
XP_016863395.1 |
XM_017007907.1 |
10639 |
Missense Mutation |
ATT,GTT |
I3445V |
XP_016863396.1 |
XM_017007908.1 |
10639 |
Missense Mutation |
ATT,GTT |
I3445V |
XP_016863397.1 |
XM_017007909.1 |
10639 |
Missense Mutation |
ATT,GTT |
I2767V |
XP_016863398.1 |
View Full Product Details