Product Details

SNP ID
rs140997717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140175265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTTCAGACCCGACCGTTATCCA[C/G]TCGGTTCGTGGAGAGGAGAGGTGAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYSTM1 PubMed Links

Gene Details

Gene
CYSTM1
Gene Name
cysteine rich transmembrane module containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032412.3 198 UTR 5 NP_115788.1
Gene
LOC101929719
Gene Name
uncharacterized LOC101929719
There are no transcripts associated with this gene.

View Full Product Details