Product Details

SNP ID
rs147102558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140114457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCCTGAAGATCGCCGAGGTGGG[C/T]GCGGGCGGCAACAAGAGCCGCCTTA
Phenotype
MIM: 600473
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01024 PubMed Links

Gene Details

Gene
LINC01024
Gene Name
long intergenic non-protein coding RNA 1024
There are no transcripts associated with this gene.

Gene
PURA
Gene Name
purine rich element binding protein A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005859.4 335 Silent Mutation GGC,GGT G92G NP_005850.1

View Full Product Details