Product Details

SNP ID
rs150900616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140632132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGCACCTGTTCCAGCCCAGCGAA[C/G]GACAGATTGAGGGAGTTCAGGGCGC
Phenotype
MIM: 158120
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD14 PubMed Links

Gene Details

Gene
CD14
Gene Name
CD14 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000591.3 1244 Silent Mutation TCC,TCG S284S NP_000582.1
NM_001040021.2 1244 Silent Mutation TCC,TCG S284S NP_001035110.1
NM_001174104.1 1244 Silent Mutation TCC,TCG S284S NP_001167575.1
NM_001174105.1 1244 Silent Mutation TCC,TCG S284S NP_001167576.1
Gene
TMCO6
Gene Name
transmembrane and coiled-coil domains 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300980.1 1244 Intron NP_001287909.1
NM_001300982.1 1244 Intron NP_001287911.1
NM_018502.4 1244 Intron NP_060972.3
XM_005268477.1 1244 Intron XP_005268534.1
XM_011537663.2 1244 Intron XP_011535965.1
XM_011537665.2 1244 Intron XP_011535967.1
XM_011537668.2 1244 Intron XP_011535970.1
XM_017009617.1 1244 Intron XP_016865106.1
XM_017009618.1 1244 Intron XP_016865107.1
XM_017009619.1 1244 Intron XP_016865108.1
XM_017009620.1 1244 Intron XP_016865109.1

View Full Product Details