Product Details

SNP ID
rs138535997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41030742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAATCTGAGGATTTCCATATAC[C/T]TGGTCTCCAAGCCCTGAGTCAACAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929555 PubMed Links

Gene Details

Gene
LOC101929555
Gene Name
uncharacterized LOC101929555
There are no transcripts associated with this gene.

Gene
UNC5CL
Gene Name
unc-5 family C-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173561.2 1232 Missense Mutation AAG,AGG K378R NP_775832.2

View Full Product Details