Product Details
- SNP ID
-
rs140821435
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:110392446 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAAAAATAGAGCTTTATGCCCTAT[A/G]CCATTAATGCTGGACTTCCTAAGTA
- Phenotype
-
MIM: 124450
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DDO
PubMed Links
Gene Details
- Gene
- DDO
- Gene Name
- D-aspartate oxidase
View Full Product Details