Product Details

SNP ID
rs142471439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29307312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACAACGTCTGCAGCATTTATCT[G/T]TTTGATCTCCATTGTGCTCTCCTAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR14J1 PubMed Links

Gene Details

Gene
OR14J1
Gene Name
olfactory receptor family 14 subfamily J member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030946.1 623 Missense Mutation TGT,TTT C208F NP_112208.1

View Full Product Details