Product Details

SNP ID
rs145423307
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110393003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCTCCCTGATGTTGCAGGCTCC[G/T]TGGAGGGAGGGCTCCAGAGCACAGC
Phenotype
MIM: 124450
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
DDO PubMed Links
Additional Information
For this assay, SNP(s) [rs35697832] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DDO
Gene Name
D-aspartate oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003649.2 709 Silent Mutation NP_003640.2
NM_004032.2 709 Silent Mutation NP_004023.2

View Full Product Details