Product Details

SNP ID
rs145654400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149400671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTTCAACAGCCTACGGGAGGT[A/G]TCTACTGAAAAGGGAACCTGCTTCT
Phenotype
MIM: 608829 MIM: 605101
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUMO4 PubMed Links

Gene Details

Gene
SUMO4
Gene Name
small ubiquitin-like modifier 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002255.1 Intron NP_001002255.1
Gene
TAB2
Gene Name
TGF-beta activated kinase 1/MAP3K7 binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292034.2 Intron NP_001278963.1
NM_001292035.2 Intron NP_001278964.1
NM_015093.5 Intron NP_055908.1
XM_011535633.2 Intron XP_011533935.1
XM_017010591.1 Intron XP_016866080.1
XM_017010592.1 Intron XP_016866081.1

View Full Product Details