Product Details

SNP ID
rs148085221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110392805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTGACTTGGGAATGGGGGTCCT[A/G]AGGGCATGGACACACTCGCTCACCA
Phenotype
MIM: 124450
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDO PubMed Links

Gene Details

Gene
DDO
Gene Name
D-aspartate oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003649.2 907 Silent Mutation CTC,CTT L360L NP_003640.2
NM_004032.2 907 Silent Mutation CTC,CTT L301L NP_004023.2

View Full Product Details