Product Details

SNP ID
rs150436455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:111566513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCTGCGACTCAGCGCCTTCCAC[A/G]TCCTGTTTGTATTTGGGGCTGATTG
Phenotype
MIM: 607043
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAF3IP2 PubMed Links

Gene Details

Gene
TRAF3IP2
Gene Name
TRAF3 interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164281.2 2637 Silent Mutation GAC,GAT D468D NP_001157753.1
NM_001164283.2 2637 Silent Mutation GAC,GAT D13D NP_001157755.1
NM_147200.2 2637 Silent Mutation GAC,GAT D478D NP_671733.2
NM_147686.3 2637 Silent Mutation GAC,GAT D469D NP_679211.2
XM_006715319.3 2637 Silent Mutation GAC,GAT D13D XP_006715382.1
XM_011535386.1 2637 Silent Mutation GAC,GAT D478D XP_011533688.1
Gene
TRAF3IP2-AS1
Gene Name
TRAF3IP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details