Product Details
- SNP ID
-
rs140584655
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:102446701 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTGAACTCCATCAGCGAGTCCCC[A/G]CATGAGCGCATGCACCCCTACATCG
- Phenotype
-
MIM: 613302
MIM: 610929
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALKBH4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1129271] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALKBH4
- Gene Name
- alkB homolog 4, lysine demethylase
There are no transcripts associated with this gene.
- Gene
- ORAI2
- Gene Name
- ORAI calcium release-activated calcium modulator 2
View Full Product Details