Product Details

SNP ID
rs141049677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30140769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTTGTATATTCTTTGAGTGGTA[G/T]TCATGAGGAGAGTAGTTACGTCAAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MTURN PubMed Links

Gene Details

Gene
MTURN
Gene Name
maturin, neural progenitor differentiation regulator homolog (Xenopus)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152793.2 Intron NP_690006.2
XM_005249652.3 Intron XP_005249709.1

View Full Product Details