Product Details
- SNP ID
-
rs144532716
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:103103479 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTATCTTGATTCTCCATGCTTCA[A/C]GACAAAAAAATCTTCAGCGTTAAGT
- Phenotype
-
MIM: 611864
MIM: 612334
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ARMC10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3181009] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARMC10
- Gene Name
- armadillo repeat containing 10
There are no transcripts associated with this gene.
- Gene
- NAPEPLD
- Gene Name
- N-acyl phosphatidylethanolamine phospholipase D
View Full Product Details