Product Details

SNP ID
rs146129746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130023607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGTCTCCTGTTCCCATGCTTCG[A/G]GTCACTGGGCGAGTTCTGGTGGTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZC3HC1 PubMed Links

Gene Details

Gene
ZC3HC1
Gene Name
zinc finger C3HC-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282190.1 1724 Silent Mutation ACC,ACT T358T NP_001269119.1
NM_001282191.1 1724 Intron NP_001269120.1
NM_016478.4 1724 Silent Mutation ACC,ACT T379T NP_057562.3
XM_005250403.2 1724 Silent Mutation ACC,ACT T255T XP_005250460.1
XM_011516288.2 1724 Silent Mutation ACC,ACT T312T XP_011514590.1
XM_011516289.2 1724 Silent Mutation ACC,ACT T289T XP_011514591.1
XM_011516290.2 1724 Silent Mutation ACC,ACT T211T XP_011514592.1
XM_017012287.1 1724 Silent Mutation ACC,ACT T336T XP_016867776.1
XM_017012288.1 1724 Silent Mutation ACC,ACT T211T XP_016867777.1

View Full Product Details