Product Details

SNP ID
rs147945300
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100064187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTACAGAACACACTTGGTGGACC[A/G]GCCCTATGACTGTAAGTGTGGAAAA
Phenotype
MIM: 194510 MIM: 601261
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF3 PubMed Links

Gene Details

Gene
ZNF3
Gene Name
zinc finger protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278284.1 1136 Intron NP_001265213.1
NM_001278287.1 1136 Intron NP_001265216.1
NM_001278290.1 1136 Intron NP_001265219.1
NM_001278291.1 1136 Intron NP_001265220.1
NM_001278292.1 1136 Intron NP_001265221.1
NM_001318135.1 1136 Intron NP_001305064.1
NM_001318136.1 1136 Intron NP_001305065.1
NM_001318137.1 1136 UTR 3 NP_001305066.1
NM_017715.3 1136 UTR 3 NP_060185.2
NM_032924.4 1136 Intron NP_116313.3
XM_006716111.1 1136 Intron XP_006716174.1
XM_017012582.1 1136 Intron XP_016868071.1
XM_017012583.1 1136 Intron XP_016868072.1
Gene
ZSCAN21
Gene Name
zinc finger and SCAN domain containing 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145914.2 1136 Missense Mutation CAG,CGG Q331R NP_666019.1
XM_005250568.4 1136 Missense Mutation CAG,CGG Q331R XP_005250625.1
XM_006716113.3 1136 Missense Mutation CAG,CGG Q331R XP_006716176.1
XM_017012584.1 1136 Missense Mutation CAG,CGG Q331R XP_016868073.1
XM_017012585.1 1136 Missense Mutation AGC,GGC S297G XP_016868074.1

View Full Product Details