Product Details

SNP ID
hCV174033290
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144432444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGTTGTAGGACAGGGACAGGGT[C/T]TTCAGGTGCTCAGCATCTGCACCGG
Phenotype
MIM: 604546 MIM: 611952
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6893 PubMed Links

Gene Details

Gene
MIR6893
Gene Name
microRNA 6893
There are no transcripts associated with this gene.

Gene
TONSL
Gene Name
tonsoku-like, DNA repair protein
There are no transcripts associated with this gene.

Gene
TONSL-AS1
Gene Name
TONSL antisense RNA 1
There are no transcripts associated with this gene.

Gene
VPS28
Gene Name
VPS28, ESCRT-I subunit
There are no transcripts associated with this gene.

View Full Product Details