Product Details

SNP ID
rs144545750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143575488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACGTCTGCTTCTCGGGGTCCTCG[A/G]TCAGCTTCATTCGTGGCTGGCCCCC
Phenotype
MIM: 130592 MIM: 611552
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EEF1D PubMed Links

Gene Details

Gene
EEF1D
Gene Name
eukaryotic translation elongation factor 1 delta
There are no transcripts associated with this gene.

Gene
MROH6
Gene Name
maestro heat like repeat family member 6
There are no transcripts associated with this gene.

Gene
NAPRT
Gene Name
nicotinate phosphoribosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286829.1 1122 Missense Mutation ACC,ATC T409I NP_001273758.1
NM_145201.5 1122 Missense Mutation ACC,ATC T409I NP_660202.3
XM_017013975.1 1122 Missense Mutation ACC,ATC T482I XP_016869464.1
XM_017013976.1 1122 Missense Mutation ACC,ATC T482I XP_016869465.1
XM_017013977.1 1122 Missense Mutation ACC,ATC T382I XP_016869466.1
XM_017013978.1 1122 Missense Mutation ACC,ATC T482I XP_016869467.1
XM_017013979.1 1122 Missense Mutation ACC,ATC T181I XP_016869468.1
XM_017013980.1 1122 Missense Mutation ACC,ATC T181I XP_016869469.1

View Full Product Details