Product Details

SNP ID
rs145057890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:52940023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCTGCCGGCGCCGCTGGCGGTGG[C/T]TGTACCAGTTGTCTACGCGGTGATC
Phenotype
MIM: 600730
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NPBWR1 PubMed Links

Gene Details

Gene
NPBWR1
Gene Name
neuropeptides B/W receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005285.3 116 Missense Mutation GCT,GTT A39V NP_005276.2

View Full Product Details