Product Details

SNP ID
rs149687434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:52940570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCCCCGTGTCCACCATCTGTGT[C/G]CTCTATACCACCCTGCTGTGCCGGC
Phenotype
MIM: 600730
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NPBWR1 PubMed Links

Gene Details

Gene
NPBWR1
Gene Name
neuropeptides B/W receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005285.3 663 Silent Mutation GTC,GTG V221V NP_005276.2

View Full Product Details