Product Details

SNP ID
rs140964923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:123034998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATAATACTCTGGTTACACCCTG[C/G]AGACTACGCCTGTGTATTTTCCTGA
Phenotype
MIM: 601909 MIM: 615882
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPR21 PubMed Links

Gene Details

Gene
GPR21
Gene Name
G protein-coupled receptor 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005294.2 7135 Missense Mutation TGC,TGG C144W NP_005285.1
XM_005251933.3 7135 Missense Mutation TGC,TGG C144W XP_005251990.1
Gene
RABGAP1
Gene Name
RAB GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012197.3 7135 Intron NP_036329.3
XM_011518440.2 7135 Intron XP_011516742.1
XM_011518441.2 7135 Intron XP_011516743.1
XM_011518442.2 7135 Intron XP_011516744.1
XM_011518444.2 7135 Intron XP_011516746.1
XM_017014567.1 7135 Intron XP_016870056.1
XM_017014568.1 7135 Intron XP_016870057.1
XM_017014569.1 7135 Intron XP_016870058.1
XM_017014570.1 7135 Intron XP_016870059.1

View Full Product Details