Product Details

SNP ID
rs143854626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92846335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGATTTCTGAGGCATACTGAGTT[A/G]TGGCTTCCAGTTTAAGGCTTTCCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD19P PubMed Links

Gene Details

Gene
ANKRD19P
Gene Name
ankyrin repeat domain 19, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF484
Gene Name
zinc finger protein 484
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007101.2 2586 Nonsense Mutation CAA,TAA Q782* NP_001007102.1
NM_001261458.1 2586 Nonsense Mutation CAA,TAA Q820* NP_001248387.1
NM_001261459.1 2586 Nonsense Mutation CAA,TAA Q782* NP_001248388.1
NM_001261460.1 2586 Nonsense Mutation CAA,TAA Q782* NP_001248389.1
NM_031486.2 2586 Nonsense Mutation CAA,TAA Q818* NP_113674.1
XM_006717299.3 2586 Nonsense Mutation CAA,TAA Q797* XP_006717362.1
XM_011519073.2 2586 Nonsense Mutation CAA,TAA Q850* XP_011517375.1
XM_011519074.2 2586 Nonsense Mutation CAA,TAA Q835* XP_011517376.1
XM_011519075.2 2586 Nonsense Mutation CAA,TAA Q833* XP_011517377.1
XM_017015178.1 2586 Nonsense Mutation CAA,TAA Q797* XP_016870667.1
XM_017015179.1 2586 Nonsense Mutation CAA,TAA Q797* XP_016870668.1
XM_017015180.1 2586 Nonsense Mutation CAA,TAA Q797* XP_016870669.1
XM_017015181.1 2586 Nonsense Mutation CAA,TAA Q782* XP_016870670.1

View Full Product Details