Product Details

SNP ID
rs144033077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37486804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGGAAGAGGAATCAACGGATC[C/T]TGGTAAGTGAAGCAGTTGCCAGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLR1E PubMed Links

Gene Details

Gene
POLR1E
Gene Name
RNA polymerase I subunit E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282766.1 291 UTR 5 NP_001269695.1
NM_022490.2 291 Silent Mutation CTG,TTG L60L NP_071935.1
XM_005251547.2 291 Missense Mutation CTG,TTG L122L XP_005251604.1

View Full Product Details