Product Details
- SNP ID
-
hCV175254336
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:154295871 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGGGTTGGACTAATGGCGGATGC[A/T]GAGGCGAGGGCTGAGTTCCCGGAGG
- Phenotype
-
MIM: 300092
MIM: 300044
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TEX28
PubMed Links
Gene Details
- Gene
- TEX28
- Gene Name
- testis expressed 28
There are no transcripts associated with this gene.
- Gene
- TKTL1
- Gene Name
- transketolase like 1
There are no transcripts associated with this gene.
View Full Product Details