Product Details

SNP ID
rs142370390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49030549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGTCGTGGCCAAGGAAAGCAGC[C/T]CTGGAGTGGGAGGTACTGGCAGGCC
Phenotype
MIM: 314310
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TFE3 PubMed Links

Gene Details

Gene
TFE3
Gene Name
transcription factor binding to IGHM enhancer 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282142.1 1628 Missense Mutation GAG,GGG E341G NP_001269071.1
NM_006521.5 1628 Missense Mutation GAG,GGG E446G NP_006512.2

View Full Product Details